Learn More
Ace Glass, Inc. Glass hose connection adapter, converts 14mm OD tube end side arm to a hose connection for 7/16in to 1/2in tubing. Order 7506-06 bushing to attach.
SDP
Supplier:  Ace Glass, Inc. 874675
Encompass_Preferred
Glass hose connection adapter, converts 14mm OD tube end side arm to a hose connection for 7/16in to 1/2in tubing. Order 7506-06 bushing to attach.
Catalog No. 50-976-329