Learn More
Ace Glass, Inc. #16 PTFE Stopper with Polypropylene Extraction Nut, Solid
SDP
Catalog No. 50977615
Encompass
#16 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-615 Supplier Ace Glass, Inc. Supplier No. 1263634
#16 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.