Learn More
Chemglass Life Sciences Stopper, Safe-Lab, Flask Length, PTFE, Size 16
SDP
Catalog No. 50872538
Encompass_Preferred
Stopper, Safe-Lab, Flask Length, PTFE, Size 16. Flask length PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the flask. All sizes are solid.
Catalog No. 50-872-538 Supplier Chemglass Life Sciences Supplier No. CG301903
Stopper, Safe-Lab, Flask Length, PTFE, Size 16. Flask length PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the flask. All sizes are solid.