Learn More
Ace Glass, Inc. #18 Ace-Thred solid PTFE plug, 7855-721 FETFE O-ring front seal
SDP
Catalog No. 50972337
Encompass_Preferred
#18 Ace-Thred solid PTFE plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -113 FETFE O-ring.
Catalog No. 50-972-337 Supplier Ace Glass, Inc. Supplier No. 584649
#18 Ace-Thred solid PTFE plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -113 FETFE O-ring.