Learn More
Ace Glass, Inc. #19 PTFE Stopper with Polypropylene Extraction Nut, Solid
SDP
Catalog No. 50983377
Encompass
#19 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-983-377 Supplier Ace Glass, Inc. Supplier No. 1263636
#19 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.