Learn More
Ace Glass, Inc. #22 PTFE Stopper with Polypropylene Extraction
SDP
Catalog No. 50977616
Encompass
#22 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-616 Supplier Ace Glass, Inc. Supplier No. 1263638
#22 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.