Learn More
Ace Glass, Inc. 10mm flash chromatography column only, epoxy coated, without disc, #25 Ace-Thred™;
SDP
Supplier:  Ace Glass, Inc. 587305
Encompass_Preferred
10mm flash chromatography column only, epoxy coated, without disc, #25 Ace-Thred™;
Catalog No. 50-972-522