Learn More
Ace Glass, Inc. #27 PTFE Stopper with Polypropylene Extraction
SDP
Catalog No. 50977617
Encompass
#27 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-617 Supplier Ace Glass, Inc. Supplier No. 1263640
#27 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.