Learn More
Chemglass Life Sciences Stopper, # 27, Polyethylene
SDP
Catalog No. CG874027
Encompass_Preferred
Stopper, # 27, Polyethylene. These blue and white stoppers are made with closed bottoms, of linear high-density polyethylene to conform to stopper dimensions. The three unit rings on the base provide an efficient seal. The improved design fully protects the neck in the event of accidental tipping. These stoppers may be used in place of the conventional flask stoppers.
Catalog No. CG874027 Supplier Chemglass Life Sciences Supplier No. CG874027
Stopper, # 27, Polyethylene. These blue and white stoppers are made with closed bottoms, of linear high-density polyethylene to conform to stopper dimensions. The three unit rings on the base provide an efficient seal. The improved design fully protects the neck in the event of accidental tipping. These stoppers may be used in place of the conventional flask stoppers.