Learn More
Ace Glass, Inc. 10.5mm ID x 25cm chromatography column, #3 stopper top, 200mL reservoir, without filter disc
SDP
Catalog No. 50972586
Encompass_Preferred
10.5mm I.D. x 25cm chromatography column with 200mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper.
Catalog No. 50-972-586 Supplier Ace Glass, Inc. Supplier No. 590705
10.5mm I.D. x 25cm chromatography column with 200mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper.