Learn More
Ace Glass, Inc. 14.5mm ID x 25cm chromatography column, #3 stopper top, 250mL reservoir, porosity B filter disc
SDP
Catalog No. 50972591
Encompass_Preferred
14.5mm I.D. x 25cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper. Includes porosity B filter disc.
Catalog No. 50-972-591 Supplier Ace Glass, Inc. Supplier No. 590735
14.5mm I.D. x 25cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper. Includes porosity B filter disc.