Learn More
Ace Glass, Inc. 30mm ID x 25cm chromatography column, #3 stopper top, 250mLreservoir, without filter disc
SDP
Catalog No. 50972588
Encompass_Preferred
30mm I.D. x 25cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper.
Catalog No. 50-972-588 Supplier Ace Glass, Inc. Supplier No. 590720
30mm I.D. x 25cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper.