Learn More
Ace Glass, Inc. 38mm ID x 30cm chromatography column, #3 stopper top, 250mLreservoir, without filter disc
SDP
Catalog No. 50972589
Encompass_Preferred
38mm I.D. x 30cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper.
Catalog No. 50-972-589 Supplier Ace Glass, Inc. Supplier No. 590722
38mm I.D. x 30cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper.