Learn More
Ace Glass, Inc. 50mm ID x 30cm chromatography column, #3 stopper top, 500mL reservoir, porosity B filter disc
SDP
Catalog No. 50978604
Encompass_Preferred
50mm I.D. x 30cm chromatography column with 500mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper. Includes porosity B filter disc.
Catalog No. 50-978-604 Supplier Ace Glass, Inc. Supplier No. 590745
50mm I.D. x 30cm chromatography column with 500mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper. Includes porosity B filter disc.