Learn More
Gilson Company Inc NO. 30 TEST SCREEN TRAY
Catalog No. NC2032711
Encompass_Preferred
No. 30 (600µm) full ASTM Testing Screen Trays feature welded and bolted heavy gauge painted steel construction and are fitted with ASTM E11 Compliance Grade wire cloth.
Catalog No. NC2032711 Supplier Gilson Company Inc Supplier No. TSA102 #30
No. 30 (600µm) full ASTM Testing Screen Trays feature welded and bolted heavy gauge painted steel construction and are fitted with ASTM E11 Compliance Grade wire cloth.