Learn More
Ace Glass, Inc. #38 PTFE Stopper with Polypropylene Extraction
SDP
Catalog No. 50983378
Encompass
#38 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-983-378 Supplier Ace Glass, Inc. Supplier No. 1263644
#38 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.