Learn More
Accuflow Systems Saint-Gobain Silicone Stopper, No. 4, Platinum Cured, Autoclavable
Catalog No. NC3314141
Encompass_Preferred
Saint-Gobain Sani-Tech Silicone Stopper No. 4 is an ultra-pure, biopharmaceutical-grade silicone closure designed for sterile laboratory and process applications. The platinum-cured formulation offers low extractables and consistent elasticity, and the stopper is compatible with steam, radiation, and gas sterilization methods.
  • Ultra-pure platinum-cured silicone for low extractables.
  • Durometer 45 Shore A for resilient sealing performance.
  • Dimensions: A 1.029 in (26.1 mm), B 0.779 in (19.8 mm), H 0.975 in (24.7 mm).
  • Sterilization: Autoclavable; radiation up to 2.5 Mrad (25 kGy); ethylene oxide compatible.
  • Tested to USP Class VI and meets FDA 177.2600 criteria.
Catalog No. NC3314141 Supplier Accuflow Systems Supplier No. 41201004
Saint-Gobain Sani-Tech Silicone Stopper No. 4 is an ultra-pure, biopharmaceutical-grade silicone closure designed for sterile laboratory and process applications. The platinum-cured formulation offers low extractables and consistent elasticity, and the stopper is compatible with steam, radiation, and gas sterilization methods.
  • Ultra-pure platinum-cured silicone for low extractables.
  • Durometer 45 Shore A for resilient sealing performance.
  • Dimensions: A 1.029 in (26.1 mm), B 0.779 in (19.8 mm), H 0.975 in (24.7 mm).
  • Sterilization: Autoclavable; radiation up to 2.5 Mrad (25 kGy); ethylene oxide compatible.
  • Tested to USP Class VI and meets FDA 177.2600 criteria.