Learn More
Ace Glass, Inc. Polypropylene #50 to 3/8" female NPT adapter with 350 micron polypropylene filter disc, retaining lock nut, and FETFE O-ring.
SDP
Supplier:  Ace Glass, Inc. 585715
Encompass_Preferred
Polypropylene #50 to 3/8" female NPT adapter with 350 micron polypropylene filter disc, retaining lock nut, and FETFE O-ring.
Catalog No. 50-972-366