Learn More
Ace Glass, Inc. #50 Ace-Thred solid nylon plug, 7855-744 FETFE O-ring front seal
SDP
Catalog No. 50972335
Encompass_Preferred
#50 Ace-Thred solid nylon plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -225 FETFE O-ring.
Catalog No. 50-972-335 Supplier Ace Glass, Inc. Supplier No. 584622
#50 Ace-Thred solid nylon plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -225 FETFE O-ring.