Learn More
Stoelting™ No. 5A Dilating Dumont INOX Forceps
SDP
Catalog No. 10001960 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
No. 5, Curved
Catalog No. Model
10-001-960 No. 5, Curved
1 options
Catalog No. 10-001-960 Supplier Stoelting™ Supplier No. 5210005A
Add to Cart
Add to Cart
Highly modified forceps have elongated, highly polished, parallel tips which are designed for controlled intraluminal vessel dilation.
Specifications
For Use With (Application) Neuroscience
Size Tip Size 0.20 X 0.16mm
Model No. 5, Curved