Learn More
Ace Glass, Inc. Offset adapter glass only, two #7 Ace-Threds on top, #15Ace-Thred bottom
SDP
Catalog No. 50971705
Encompass_Preferred
Offset adapter glass only, two #7 Ace-Threds on top, #15Ace-Thred bottom
Catalog No. 50-971-705 Supplier Ace Glass, Inc. Supplier No. 510205
Offset adapter glass only, two #7 Ace-Threds on top, #15Ace-Thred bottom