Learn More
Ace Glass, Inc. #7-#15 feed tube with hose connection
SDP
Catalog No. 50972218
Encompass_Preferred
Feed tube adapter with #7 to #15 Ace-Thred ends and hose connection on side arm. #7 thread at top is supplied with a nylon bushing and FETFE o-ring, for use with adjustable feed tubes.
Catalog No. 50-972-218 Supplier Ace Glass, Inc. Supplier No. 582724
Feed tube adapter with #7 to #15 Ace-Thred ends and hose connection on side arm. #7 thread at top is supplied with a nylon bushing and FETFE o-ring, for use with adjustable feed tubes.