Learn More
Ace Glass, Inc. #7-#50 feed tube with hose connection
SDP
Catalog No. 50972219
Encompass_Preferred
Feed tube adapter with #7 to #50 Ace-Thred ends and hose connection on side arm. #7 thread at top is supplied with a nylon bushing and FETFE o-ring, for use with adjustable feed tubes.
Catalog No. 50-972-219 Supplier Ace Glass, Inc. Supplier No. 582730
Feed tube adapter with #7 to #50 Ace-Thred ends and hose connection on side arm. #7 thread at top is supplied with a nylon bushing and FETFE o-ring, for use with adjustable feed tubes.