Learn More
Ace Glass, Inc. Coupling, #7-#7, with ferrule for tubing attachment, connects solvent reservoir to washer body
SDP
Catalog No. 50971329
Encompass_Preferred
#7 Ace-Thred to #7 Ace-Thred coupling connects the solvent reservoir to the NMR tube washer body. Coupling has a ferrule for tubing attachment.
Catalog No. 50-971-329 Supplier Ace Glass, Inc. Supplier No. 254019
#7 Ace-Thred to #7 Ace-Thred coupling connects the solvent reservoir to the NMR tube washer body. Coupling has a ferrule for tubing attachment.