Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC No-711 hydrochloride | 145645-62-1 | MFCD00153853 | 99.8% | 386.87 g/mol | C21H23ClN2O3 | 100 MG
SDP

Catalog No. 5000248162
Encompass_Preferred

NNC-711 (NO-711 hydrochloride) is a research-grade GAT-1 (GABA transporter 1) inhibitor supplied as the hydrochloride salt for laboratory assays. It is intended for in vitro and in vivo research use to block GABA reuptake in neuropharmacology studies.

  • Potent, selective GAT-1 inhibitor used in neuroscience research.
  • Cas number 145645-62-1.
  • Molecular formula C21H23ClN2O3; molecular weight 386.87 g/mol.
  • Purity approximately 99.8% as supplied.
  • Form: hydrochloride salt; appearance: white to off-white solid.
  • Storage: keep sealed and dry; store at -20°C (dry) or -80°C in solvent for long-term.
  • Typical pack size: 100 MG.

Catalog No. 50-002-48162 Supplier Medchemexpress LLC Supplier No. HY103506100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NNC-711 (NO-711 hydrochloride) is a research-grade GAT-1 (GABA transporter 1) inhibitor supplied as the hydrochloride salt for laboratory assays. It is intended for in vitro and in vivo research use to block GABA reuptake in neuropharmacology studies.

  • Potent, selective GAT-1 inhibitor used in neuroscience research.
  • Cas number 145645-62-1.
  • Molecular formula C21H23ClN2O3; molecular weight 386.87 g/mol.
  • Purity approximately 99.8% as supplied.
  • Form: hydrochloride salt; appearance: white to off-white solid.
  • Storage: keep sealed and dry; store at -20°C (dry) or -80°C in solvent for long-term.
  • Typical pack size: 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.