Learn More
Ace Glass, Inc. Column, 8" ID x 18" , 6" O-ring grooved flat flange top, #80 Ace-Thred bottom
SDP
Catalog No. 50978536
Encompass_Preferred
8 I.D. x 18 chromatography column with a size 6 o-ring grooved flat flange top and #80 Ace-Thred bottom. Supplied with silicone o-ring.
Catalog No. 50-978-536 Supplier Ace Glass, Inc. Supplier No. 586218
8 I.D. x 18 chromatography column with a size 6 o-ring grooved flat flange top and #80 Ace-Thred bottom. Supplied with silicone o-ring.