Learn More
Ace Glass, Inc. Column, 8" ID x 24" , 6" O-ring grooved flat flange top, #80 Ace-Thred bottom
SDP
Catalog No. 50972390
Encompass_Preferred
8 I.D. x 24 chromatography column with a size 6 o-ring grooved flat flange top and #80 Ace-Thred bottom. Supplied with silicone o-ring.
Catalog No. 50-972-390 Supplier Ace Glass, Inc. Supplier No. 586220
8 I.D. x 24 chromatography column with a size 6 o-ring grooved flat flange top and #80 Ace-Thred bottom. Supplied with silicone o-ring.