Learn More
Ace Glass, Inc. Large column stand set, up to #80 thread
SDP
Catalog No. 50977155
Encompass
#80 Ace-Thred Support Stand includes U-shaped base, two support rods, 5/8 diameter x 29 long, and one rod 5/8x 5 long. Also includes 3 power hold clamps, 3 Chain Clamps, a modified Alum Support Stand, and one Cork Ring.
Catalog No. 50-977-155 Supplier Ace Glass, Inc. Supplier No. 1105635
#80 Ace-Thred Support Stand includes U-shaped base, two support rods, 5/8 diameter x 29 long, and one rod 5/8x 5 long. Also includes 3 power hold clamps, 3 Chain Clamps, a modified Alum Support Stand, and one Cork Ring.