Learn More
Ace Glass, Inc. #80 Ace-Thred solid PTFE plug, 7855-764 FETFE O-ring front seal
SDP
Catalog No. 50972338
Encompass_Preferred
#80 Ace-Thred solid PTFE plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -235 FETFE O-ring.
Catalog No. 50-972-338 Supplier Ace Glass, Inc. Supplier No. 584660
#80 Ace-Thred solid PTFE plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -235 FETFE O-ring.