Learn More
Ace Glass, Inc. #80 Ace-Thred solid UHMWP plug, 7855-764 FETFE O-ring front seal
SDP
Catalog No. 50978518
Encompass_Preferred
#80 Ace-Thred solid nylon plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -235 FETFE O-ring.
Catalog No. 50-978-518 Supplier Ace Glass, Inc. Supplier No. 584627
#80 Ace-Thred solid nylon plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -235 FETFE O-ring.