Learn More
Ace Glass, Inc. #9 PTFE Stopper with Polypropylene Extraction
SDP
Catalog No. 50977613
Encompass
#9 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-613 Supplier Ace Glass, Inc. Supplier No. 1263630
#9 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.