Learn More
Enzo Life Sciences ROS-ID NO Detection kit (1x1Kit)
SDP
Catalog No. NC0870786
Encompass_Preferred
Applications: Fluorescence microscopy. Fluorescent detection. Contents: NO Detection Reagent (Red), 60 µl. NO Inducer (L-Arginine), 100 µl. NO Scavenger (c-PTIO), 400 nmoles. 10X Wash Buffer, 15 ml
Catalog No. NC0870786 Supplier Enzo Life Sciences Supplier No. ENZ51013200
Applications: Fluorescence microscopy. Fluorescent detection. Contents: NO Detection Reagent (Red), 60 µl. NO Inducer (L-Arginine), 100 µl. NO Scavenger (c-PTIO), 400 nmoles. 10X Wash Buffer, 15 ml