Learn More
Anton Paar No need for pressurized air; place it wherever you need it Measurements in rotation and special-application oscillation Ideal for measuring sample flow behavior
Catalog No. 5000580502
Encompass_Preferred
MCR 53 SpecificationsBearing design: MechanicalMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: n/aMin. torque (rotation): 200 µNmMin. torque (oscillation): 200 µNmMax. torque: 125 mNmTorque resolution: 5 nNmAngular deflection resolution: 8 nradMin. angular velocity: 10-8 rad/sMax. angular velocity / max. speed: 157 rad/s (1,500 rpm)Min. frequency: 10-7 HzMax. frequency: 100 HzNormal force range: n/aNormal force resolution: 0.4 mNAuto Control: YesTruStrain: n/aTruRate: n/a
Catalog No. 50-005-80502 Supplier Anton Paar Supplier No. 385252
Please contact your sales representative for a quote or further ordering instructions.
MCR 53 SpecificationsBearing design: MechanicalMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: n/aMin. torque (rotation): 200 µNmMin. torque (oscillation): 200 µNmMax. torque: 125 mNmTorque resolution: 5 nNmAngular deflection resolution: 8 nradMin. angular velocity: 10-8 rad/sMax. angular velocity / max. speed: 157 rad/s (1,500 rpm)Min. frequency: 10-7 HzMax. frequency: 100 HzNormal force range: n/aNormal force resolution: 0.4 mNAuto Control: YesTruStrain: n/aTruRate: n/a