Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec No Sink Countertop Trimming Station
SDP

Catalog No. 22444225 Shop All Mopec Products
Change view
Click to view available options
Height (Metric):
116 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
22-444-225 116 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-225 Supplier Mopec Supplier No. OB100

missing translation for 'validMainframeMsg'

Ideal for laboratories with space constraints and can serve as a secondary station in larger, busier, facilities that require additional work space. Mopec™ No Sink Countertop Trimming Table is a compact workstation that rests on a countertop and saves valuable floor space.

  • Includes hot/cold water fixture, wrist blade handles with swing spout faucet and built-in vacuum breaker
  • Recessed stainless steel work surface
  • Washable ruler with inch/centimeter markings
  • Ruler is located on the inside lip of the workstation
  • Magnetic tool bar holds instruments when not in use
  • One large and one small stainless steel organizer bin
  • Four leveling pads
  • Stainless steel c-fold paper towel dispenser
  • Two energy saving, long life LED lights
  • GFCI Electrical Receptacle, 1 circuit – 15 amps
  • 8 foot hospital grade line cord
  • Back draft ventilation
  • Designed to safely and efficiently remove formaldehyde fumes and hazardous chemical vapors
  • Equipped with a factory installed exhaust duct which can be connected to any in-house ventilation system
  • All plumbing and electrical work accomplished at factory
  • Only trade hookup required

Warranty and Services

1 Year Warranty

Specifications

Product Type Trimming Station
Includes Removable polyethylene cutting board 58.4 x 40.6 x 2cm
Frequency 60 Hz
Voltage 115 V
Depth (English) 48 in.
Depth (English) Sink 3 in.
Depth (Metric) 122 cm
Depth (Metric) Sink 7.6 cm
Height (English) 45.5 in.
Width (English) 29 in.
Height (Metric) 116 cm
Length (English) Sink 10 in.
Length (Metric) Sink 25.4cm
Width (English) Sink 5 in.
Width (Metric) 74 cm
Width (Metric) Sink 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.