Learn More
IHEALTH LABS INC iHealth Non-Contact Infrared Forehead Thermometer
SDP
Catalog No. NC1810996
Encompass_Preferred
iHealth Thermometer PT3 is a non-contact infrared forehead thermometer that displays body temperature readings within 1 second. With a built-in distance sensor and environmental sensor, our infrared digital thermometer reduces the chance of invalid measurements and makes adjustments for accurate readings.
This item is not returnable. View return policy
Catalog No. NC1810996 Supplier IHEALTH LABS INC Supplier No. PT3
iHealth Thermometer PT3 is a non-contact infrared forehead thermometer that displays body temperature readings within 1 second. With a built-in distance sensor and environmental sensor, our infrared digital thermometer reduces the chance of invalid measurements and makes adjustments for accurate readings.