Learn More
Poltex No Waste Glove Box Holder with Stand
SDP
Catalog No. 22000351
Change view
Click to view available options
Capacity:
3 Boxes
4 Boxes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
22-000-351 4 Boxes
22-000-350 3 Boxes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-000-351 Supplier Poltex Supplier No. NOWASTGB43STL
  • $981.00
    $765.65 / Each of 1
    Save $215.35
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Unique design positions gloves directly facing up and features bottleneck slots to prevent clumps from dispensing, only single gloves. Glove boxes are secured in place with a sliding door and padlock (included).
  • Unique design positions gloves directly facing up
  • Bottleneck slots prevent clumps from dispensing, only single gloves
  • Glove box holder is constructed of durable PETG with a powder-coated steel stand
  • Boxes are secured in place with a sliding door and padlock (included)
  • Steel stand can accept additional magnet-mounted accessories such as wipe canisters or masks (sold separately)
Specifications
Product Type Glove Box Holder
Color Clear, Gray
Dimensions (D x W x H) Interior 10.75 x 22 x 3.5 in.
Holds Disposable Glove Boxes
Material PETG
Mounting Stand Mount
No. of Compartments 4
Product Style Top Loading
Capacity 4 Boxes
Includes Padlock