Learn More
Brookfield Engineer Labs NO1 SPINDLE BROOKFIELD RV
Catalog No. NC9128518
Encompass
Spindle, Entry Code: 01, SMC: 01, SRC: 0, Each spindle has a two digit entry code which is entered via the keypad on the DV-II +, The entry codeallows the DV-II + to calculate Viscosity, Shear Rate and Shear Stress values.
Catalog No. NC9128518 Supplier Brookfield Engineer Labs Supplier No. RV1
Spindle, Entry Code: 01, SMC: 01, SRC: 0, Each spindle has a two digit entry code which is entered via the keypad on the DV-II +, The entry codeallows the DV-II + to calculate Viscosity, Shear Rate and Shear Stress values.