Learn More
Chemglass Life Sciences Chem-Thread, Size 12, 17mm Tube OD, Accepts 9-12mm OD Tubing, #205 O-ring
SDP
Catalog No. CG35006
Encompass_Preferred
Chem-Thread, Size 12, 17mm Tube OD, Accepts 9-12mm OD Tubing, #205 O-ring. Consists of an externally threaded glass tube, Viton o-ring and Celcon compression cap. Tube O.D. listed is the shank O.D. of the connector.
Catalog No. CG35006 Supplier Chemglass Life Sciences Supplier No. CG35006
Chem-Thread, Size 12, 17mm Tube OD, Accepts 9-12mm OD Tubing, #205 O-ring. Consists of an externally threaded glass tube, Viton o-ring and Celcon compression cap. Tube O.D. listed is the shank O.D. of the connector.