Learn More
Chemglass Life Sciences Stopper, Flask Length, Flathead, Solid, Size 13
SDP
Catalog No. CG301601
Encompass_Preferred
Stopper, Flask Length, Flathead, Solid, Size 13. Flathead design allows stopper to be inverted and placed on end thereby preventing contamination of the ground surface.
Catalog No. CG301601 Supplier Chemglass Life Sciences Supplier No. CG301601
Stopper, Flask Length, Flathead, Solid, Size 13. Flathead design allows stopper to be inverted and placed on end thereby preventing contamination of the ground surface.