Learn More
Chemglass Life Sciences Loosening Ring, Rodaviss, Size 14
SDP
Catalog No. CG18401
Encompass_Preferred
Loosening Ring, Rodaviss, Size 14. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.
Catalog No. CG18401 Supplier Chemglass Life Sciences Supplier No. CG18401
Loosening Ring, Rodaviss, Size 14. Split Polyamide loosening ring to enable inner and outer Rodaviss members to be pulled apart in the event of freezing.