Learn More
Chemglass Life Sciences Silicone Sealing Ring, fits GL-14 Cap, fits tubing O.D. 5.5mm to 6.5mm
SDP
Catalog No. CG19701
Encompass_Preferred
Silicone Sealing Ring, fits GL-14 Cap, fits tubing O.D. 5.5mm to 6.5mm. Seal is made from silicone with a vulcanized PTFE washer having an aperture to fit the listed tubing size. For use with CG-195 cap.
Catalog No. CG19701 Supplier Chemglass Life Sciences Supplier No. CG19701
Silicone Sealing Ring, fits GL-14 Cap, fits tubing O.D. 5.5mm to 6.5mm. Seal is made from silicone with a vulcanized PTFE washer having an aperture to fit the listed tubing size. For use with CG-195 cap.