Learn More
Ace Glass, Inc. Tube only, pressure, 140mL, #15, 38.1mm OD, 20.3cm long, no plug
SDP
Catalog No. NC9499763
Encompass_Preferred
120mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 38.1mm O.D. x 20.3cm long.
Catalog No. NC9499763 Supplier Ace Glass, Inc. Supplier No. 864830
120mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 38.1mm O.D. x 20.3cm long.