Learn More
Chemglass Life Sciences Stopper, Flask Length, Flathead, Solid, Size 16
SDP
Catalog No. CG301602
Encompass_Preferred
Stopper, Flask Length, Flathead, Solid, Size 16. Flathead design allows stopper to be inverted and placed on end thereby preventing contamination of the ground surface.
Catalog No. CG301602 Supplier Chemglass Life Sciences Supplier No. CG301602
Stopper, Flask Length, Flathead, Solid, Size 16. Flathead design allows stopper to be inverted and placed on end thereby preventing contamination of the ground surface.