Learn More
Cole-Parmer Cole-Parmer Solid Black Rubber Stoppers, Standard Size 2; 52/Pk
Catalog No. NC9467976 Shop All Cole-Parmer Products
Encompass_Preferred
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.
Catalog No. NC9467976 Supplier Cole-Parmer Supplier No. 6299008
Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.