Learn More
Chemglass Life Sciences Cap w/hole, SVL, Size 22, Fits Tubing 15.60-16.40mm OD
SDP
Catalog No. CG18903
Encompass_Preferred
Cap w/hole, SVL, Size 22, Fits Tubing 15.60-16.40mm OD. Open top SVL screw cap supplied complete with PTFE sealing ring to accept the tubing size as listed.
Catalog No. CG18903 Supplier Chemglass Life Sciences Supplier No. CG18903
Cap w/hole, SVL, Size 22, Fits Tubing 15.60-16.40mm OD. Open top SVL screw cap supplied complete with PTFE sealing ring to accept the tubing size as listed.