Learn More
Chemglass Life Sciences Stopper, Safe-Lab, Flask Length, PTFE, Size 22
SDP
Catalog No. 50872540
Encompass_Preferred
Stopper, Safe-Lab, Flask Length, PTFE, Size 22. Flask length PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the flask. All sizes are solid.
Catalog No. 50-872-540 Supplier Chemglass Life Sciences Supplier No. CG301905
Stopper, Safe-Lab, Flask Length, PTFE, Size 22. Flask length PTFE stopper has a Polypropylene extraction nut permitting easy removal of the stopper from the flask. All sizes are solid.