Learn More
Chemglass Life Sciences Screw Thread Tube, SVL, Size 22, on 22mm OD x 1.8mm W tubing, Approx. 150mm OAL
SDP
Catalog No. CG18702
Encompass_Preferred
Screw Thread Tube, SVL, Size 22, on 22mm OD x 1.8mm W tubing, 165mm OAL. Screw thread tube for replacement of apparatus using the SVL series of glass threads. For use with a CG-188 solid cap with PTFE liner or a CG-189 open top cap and sealing ring for use as a tubing connector. Threads are tooled on one end of a 6in length of tubing.
Catalog No. CG18702 Supplier Chemglass Life Sciences Supplier No. CG18702
Screw Thread Tube, SVL, Size 22, on 22mm OD x 1.8mm W tubing, 165mm OAL. Screw thread tube for replacement of apparatus using the SVL series of glass threads. For use with a CG-188 solid cap with PTFE liner or a CG-189 open top cap and sealing ring for use as a tubing connector. Threads are tooled on one end of a 6in length of tubing.