Learn More
Chemglass Life Sciences Cap, #24, With Hole, Rodaviss
SDP
Catalog No. CG18203
Encompass_Preferred
Cap, #24, With Hole, Rodaviss. Threaded cap fits CG-172 outer joint, Rodaviss. When used with CG-305 Viton o-rings, provides for greaseless high vacuum seal.
Catalog No. CG18203 Supplier Chemglass Life Sciences Supplier No. CG18203
Cap, #24, With Hole, Rodaviss. Threaded cap fits CG-172 outer joint, Rodaviss. When used with CG-305 Viton o-rings, provides for greaseless high vacuum seal.