Learn More
Ace Glass, Inc. Bottle only, pressure, 75mL, #25, no plug
SDP
Catalog No. 509558520
Encompass_Preferred
75mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Does not include plug.
Catalog No. 50-955-8520 Supplier Ace Glass, Inc. Supplier No. 8648135
75mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Does not include plug.