Learn More
Ace Glass, Inc. Bottle only, pressure, 175mL, #25, no plug
SDP
Catalog No. 509558521
Encompass_Preferred
175mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Does not include #25 plug.
Catalog No. 50-955-8521 Supplier Ace Glass, Inc. Supplier No. 8648138
175mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Does not include #25 plug.